gRNA Design Details
- Target DNA Sequence
- AAAAGCGTTCGGGGTGACTG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 206476722 - 206476744 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IKBKE
- Entrez Gene ID
-
9641
- Gene Name
- inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon
- Gene Aliases
-
IKK-E, IKK-i, IKKE, IKKI
- Chromosome Location
-
Chr.1: 206470243 - 206496890 on GRCh38
- UniGene ID
- Hs.321045
- Species
-
Human