gRNA Design Details
- Target DNA Sequence
- AGAATCGCACAGTCTCTGGT
- PAM Sequence
- GGG
- Target locus
-
Chr.12: 53382360 - 53382382 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.83/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SP1
- Entrez Gene ID
-
6667
- Gene Name
- Sp1 transcription factor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 53380195 - 53416446 on GRCh38
- UniGene ID
- Hs.620754
- Species
-
Human