gRNA Design Details

Target DNA Sequence
TGTAGGAGTGATTGTCCTCA
PAM Sequence
AGG
Target locus
Chr.20: 62809634 - 62809656 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 87.21/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
OGFR
Entrez Gene ID
11054
Gene Name
opioid growth factor receptor
Gene Aliases
-
Chromosome Location
Chr.20: 62804825 - 62814000 on GRCh38
UniGene ID
Hs.67896
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_007346.2 NP_031372.2 4