gRNA Design Details

Target DNA Sequence
AATGCCAATGCCAGGGCAAA
PAM Sequence
GGG
Target locus
Chr.20: 35437696 - 35437718 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 85.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GDF5
Entrez Gene ID
8200
Gene Name
growth differentiation factor 5
Gene Aliases
BDA1C, BMP-14, BMP14, CDMP1, LAP-4, LAP4, OS5, SYM1B, SYNS2
Chromosome Location
Chr.20: 35433347 - 35454746 on GRCh38
UniGene ID
Hs.1573
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000557.4 NP_000548.2 1
NM_001319138.1 NP_001306067.1 3
XM_011529075.2 XP_011527377.1 3