gRNA Design Details
- Target DNA Sequence
- ACATTAGTGCTACGCGGAAT
- PAM Sequence
- GGG
- Target locus
-
Chr.19: 41238491 - 41238513 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
96.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AXL
- Entrez Gene ID
-
558
- Gene Name
- AXL receptor tyrosine kinase
- Gene Aliases
-
ARK, JTK11, Tyro7, UFO
- Chromosome Location
-
Chr.19: 41219199 - 41261767 on GRCh38
- UniGene ID
- Hs.590970
- Species
-
Human