gRNA Design Details
- Target DNA Sequence
- CCTTGTACTTGGGCTCGTTC
- PAM Sequence
- CGG
- Target locus
-
Chr.17: 81688769 - 81688791 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HGS
- Entrez Gene ID
-
9146
- Gene Name
- hepatocyte growth factor-regulated tyrosine kinase substrate
- Gene Aliases
-
HRS
- Chromosome Location
-
Chr.17: 81683932 - 81702121 on GRCh38
- UniGene ID
- Hs.661056
- Species
-
Human