gRNA Design Details

Target DNA Sequence
AGTCGATGTTGGAGCCAGTG
PAM Sequence
CGG
Target locus
Chr.1: 15506970 - 15506992 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 92.63/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CASP9
Entrez Gene ID
842
Gene Name
caspase 9
Gene Aliases
APAF-3, APAF3, ICE-LAP6, MCH6, PPP1R56
Chromosome Location
Chr.1: 15491401 - 15524912 on GRCh38
UniGene ID
Hs.329502
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001229.4 NP_001220.2 4
NM_032996.3 NP_127463.2 4
NR_102732.1 4
NR_102733.1 4
XM_005246014.2 XP_005246071.1 4
XM_011542273.2 XP_011540575.1 4