Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320653.1 | NP_001307582.1 | 3 | 612 | |
NM_002532.5 | NP_002523.2 | 3 | 612 | ||
XM_011523893.2 | XP_011522195.1 | 4 | 662 | ||
XM_017024694.1 | XP_016880183.1 | 4 | 662 | ||
GenBank mRNA | AK225247.1 | - | 3 | 535 | |
AK298158.1 | - | 4 | 389 | ||
AK299000.1 | - | 3 | 535 | ||
AK308142.1 | - | 3 | 424 | ||
BC000335.2 | - | 3 | 525 | 2822595 | |
Y08612.2 | - | 3 | 571 |