Gene Transcripts
- Gene Symbol
- IL1R2
- Entrez Gene ID
- 7850
- Gene Name
- interleukin 1 receptor type 2
- Gene Aliases
- CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB
- Chromosome Location
- Chr. 2: 101991805 - 102028544 on Build GRCh38
- UniGene ID
- Hs.25333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001261419.1 |
NP_001248348.1
|
6, 7 |
991 |
|
NM_004633.3 |
NP_004624.1
|
6, 7 |
991 |
|
NR_048564.1 |
|
6, 7 |
978 |
|
XM_006712734.3 |
XP_006712797.1
|
6, 7 |
1015 |
|
XM_006712736.3 |
XP_006712799.1
|
6, 7 |
1052 |
|
XM_011511801.2 |
XP_011510103.1
|
6, 7 |
3351 |
|
XM_011511803.2 |
XP_011510105.1
|
6, 7 |
2930 |
|
XM_011511804.2 |
XP_011510106.1
|
6, 7 |
1044 |
|
XM_011511805.2 |
XP_011510107.1
|
6, 7 |
1817 |
|
XM_011511808.2 |
XP_011510110.1
|
2, 3 |
646 |
|
XM_017004889.1 |
XP_016860378.1
|
5, 6 |
901 |
|
GenBank mRNA |
AK130412.1 |
- |
6, 7 |
875 |
|
BC039031.1 |
- |
6, 7 |
991 |
5723005 |
FJ200480.1 |
- |
5, 6 |
602 |
|
U64094.1 |
- |
5, 6 |
823 |
|
U74649.1 |
- |
5, 6 |
797 |
|
X59770.1 |
- |
5, 6 |
823 |
|
View Full Product Details