Gene Transcripts

Gene Symbol
RSPRY1
Entrez Gene ID
89970
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
SEMDFA
Chromosome Location
Chr. 16: 57186284 - 57240475 on Build GRCh38
UniGene ID
Hs.460885
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305163.1 NP_001292092.1 14 2509
NM_001305164.1 NP_001292093.1 14 1888
NM_133368.2 NP_588609.1 14 1910
XM_005256220.1 XP_005256277.1 14 2217
XM_011523427.1 XP_011521729.1 15 1996
XM_011523428.1 XP_011521730.1 15 2310
XM_011523430.1 XP_011521732.1 15 1937
XM_017023844.1 XP_016879333.1 15 2683
GenBank mRNA AB075852.1 - 14 1830
AK027549.1 - 14 2460
AK124601.1 - 6 2827
AK172845.1 - 14 1866
AL834402.1 - 2 114
BC013173.2 - 14 1865 4340287

View Full Product Details