Gene Transcripts

Gene Symbol
MRGPRD
Entrez Gene ID
116512
Gene Name
MAS related GPR family member D
Gene Aliases
MRGD, TGR7
Chromosome Location
Chr. 11: 68980021 - 68980986 on Build GRCh38
UniGene ID
Hs.527802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198923.2 NP_944605.2 1 37
GenBank mRNA AB154410.1 - 1 37
AY427820.1 - 1 37

View Full Product Details