Gene Transcripts

Gene Symbol
TMEM86A
Entrez Gene ID
144110
Gene Name
transmembrane protein 86A
Gene Aliases
Chromosome Location
Chr. 11: 18698735 - 18704785 on Build GRCh38
UniGene ID
Hs.502100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153347.2 NP_699178.1 2 281
GenBank mRNA AK074600.1 - 2 237
BC017073.2 - 1 69
BC035692.1 - 2 212 5759114
DA047874.1 - 2 281

View Full Product Details