Gene Transcripts

Gene Symbol
OFCC1
Entrez Gene ID
266553
Gene Name
orofacial cleft 1 candidate 1
Gene Aliases
MRDS1
Chromosome Location
Chr. 6: 9775808 - 9842630 on Build GRCh38
UniGene ID
Hs.532138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF520800.1 - 4, 5 355
AF520803.1 - 2, 3 227
AF520804.1 - 2, 3 213
AF520805.1 - 2, 3 227
AF520806.1 - 2, 3 235
AF520807.1 - 3, 4 285
AF520808.1 - 3, 4 315
AF548113.1 - 4, 5 501
AF548114.1 - 140
AF548116.1 - 1 12
AF548117.1 - 135
AF548118.1 - 188
AF548122.1 - 1, 2 188
AF548123.1 - 1, 2 188
AY309094.1 - 5, 6 436
BC113541.1 - 3, 4 294 8322593
BC113543.1 - 3, 4 294 8322595
BC143837.1 - 4, 5 306 9052352

Gene Symbol
LOC107983965
Entrez Gene ID
107983965
Gene Name
orofacial cleft 1 candidate gene 1 protein-like
Gene Aliases
Chromosome Location
Chr. 6: 9850416 - 10211608 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017011612.1 XP_016867101.1 5, 6 652

View Full Product Details