Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
Chromosome Location
Chr. 11: 64606174 - 64723188 on Build GRCh38
UniGene ID
Hs.372938
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015080.3 NP_055895.1 21 4404
NM_138732.2 NP_620060.1 18 4194
NM_138734.2 NP_620063.1 5 1316
XM_005274400.3 XP_005274457.1 20 3936
XM_005274401.3 XP_005274458.1 20 3936
XM_005274402.3 XP_005274459.1 19 3912
XM_011545370.1 XP_011543672.1 20 3966
XM_011545371.1 XP_011543673.1 20 3963
XM_011545373.1 XP_011543675.1 20 3945
XM_011545375.1 XP_011543677.1 19 3921
XM_011545385.1 XP_011543687.1 13 2311
XM_017018563.1 XP_016874052.1 19 3936
XM_017018564.1 XP_016874053.1 19 3933
XM_017018565.1 XP_016874054.1 19 3930
XM_017018566.1 XP_016874055.1 19 3894
XM_017018567.1 XP_016874056.1 19 3876
XM_017018568.1 XP_016874057.1 19 3867
XM_017018569.1 XP_016874058.1 18 3864
XM_017018570.1 XP_016874059.1 20 3966
XM_017018571.1 XP_016874060.1 20 3957
XM_017018572.1 XP_016874061.1 20 3261
XM_017018573.1 XP_016874062.1 18 3840
XM_017018574.1 XP_016874063.1 19 3231
XM_017018575.1 XP_016874064.1 19 3228
XM_017018576.1 XP_016874065.1 15 2876
XM_017018577.1 XP_016874066.1 13 2302
XM_017018578.1 XP_016874067.1 19 3867
GenBank mRNA AB023138.2 - 19 4171
AB035266.1 - 20 4056
AB177862.1 - 17 3108
AK098211.1 - 1 1013
BC150275.1 - 19 4171 9007249

View Full Product Details