Gene Transcripts

Gene Symbol
NFAM1
Entrez Gene ID
150372
Gene Name
NFAT activating protein with ITAM motif 1
Gene Aliases
CNAIP
Chromosome Location
Chr. 22: 42380407 - 42438155 on Build GRCh38
UniGene ID
Hs.436677
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318323.1 NP_001305252.1 4, 5 681
NM_145912.6 NP_666017.1 5, 6 794
XM_005261374.4 XP_005261431.1 7, 8 1421
GenBank mRNA AK074094.1 - 4, 5 386
AY121370.1 - 5, 6 794
AY247409.1 - 5, 6 789
BC038241.1 - 5, 6 789 5180790
BI906855.1 - 4, 5 655
CT841509.1 - 5, 6 794

View Full Product Details