Gene Transcripts

Gene Symbol
NHSL2
Entrez Gene ID
340527
Gene Name
NHS like 2
Gene Aliases
Chromosome Location
Chr. X: 71911073 - 72153286 on Build GRCh38
UniGene ID
Hs.397836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013627.2 NP_001013649.2 6 1486
XM_011530932.1 XP_011529234.1 7 1570
XM_011530933.1 XP_011529235.1 6 1438
XM_011530934.2 XP_011529236.1 6 6278
XM_017029473.1 XP_016884962.1 6 5248
XM_017029474.1 XP_016884963.1 6 1390
XM_017029475.1 XP_016884964.1 2 4633
XM_017029476.1 XP_016884965.1 2 750
XM_017029477.1 XP_016884966.1 2 5664
GenBank mRNA BC033261.1 - 2 1650
BC136756.1 - 3 859 9020746

View Full Product Details