Gene Transcripts
- Gene Symbol
- NKX2-4
- Entrez Gene ID
- 644524
- Gene Name
- NK2 homeobox 4
- Gene Aliases
- NKX2.4, NKX2D
- Chromosome Location
- Chr. 20: 21395367 - 21397409 on Build GRCh38
- UniGene ID
- Hs.456662
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033176.1 |
NP_149416.1
|
1 |
13 |
|
View Full Product Details