Gene Transcripts

Gene Symbol
CLPTM1
Entrez Gene ID
1209
Gene Name
CLPTM1, transmembrane protein
Gene Aliases
Chromosome Location
Chr. 19: 44954585 - 44993346 on Build GRCh38
UniGene ID
Hs.444441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294.3 NP_001285.1 1, 2 242
GenBank mRNA AF037339.1 - 1, 2 85
AK074935.1 - 1, 2 86
AK075262.1 - 1, 2 85
AK223553.1 - 1, 2 86
BC004865.2 - 1, 2 121 3840991
BC012359.1 - 1, 2 75 4685026
BI916245.1 - 1, 2 88
BT007262.1 - 1, 2 70
DB033341.1 - 1, 2 220
HY033360.1 - 1, 2 243
KU177986.1 - 1, 2 70
KU177987.1 - 1, 2 70

View Full Product Details