Gene Transcripts

Gene Symbol
EDN2
Entrez Gene ID
1907
Gene Name
endothelin 2
Gene Aliases
ET-2, ET2, PPET2
Chromosome Location
Chr. 1: 41478775 - 41484699 on Build GRCh38
UniGene ID
Hs.1407
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302269.1 NP_001289198.1 3 315
NM_001956.4 NP_001947.1 3 315
NR_126098.1 3 224
GenBank mRNA BC034393.1 - 3 258
BI761655.1 - 3 214
HY177176.1 - 3 316
M65199.1 - 3 303
S63833.1 - 4 270
S63834.1 - 2 51
S63836.1 - 2 51
S63837.1 - 2 51
S63838.1 - 3 103
X55177.1 - 3 291

View Full Product Details