Gene Transcripts

Gene Symbol
WDFY3
Entrez Gene ID
23001
Gene Name
WD repeat and FYVE domain containing 3
Gene Aliases
ALFY, ZFYVE25
Chromosome Location
Chr. 4: 84669537 - 84966391 on Build GRCh38
UniGene ID
Hs.480116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014991.4 NP_055806.2 40 6960
XM_005262858.4 XP_005262915.1 42 7001
XM_011531757.2 XP_011530059.1 41 6912
XM_011531759.2 XP_011530061.1 40 6874
XM_011531760.2 XP_011530062.1 40 6812
XM_011531761.2 XP_011530063.1 41 6867
XM_011531762.2 XP_011530064.1 41 6912
XM_011531763.2 XP_011530065.1 39 6758
XM_011531764.2 XP_011530066.1 40 6828
XM_011531765.2 XP_011530067.1 38 6614
XM_011531766.2 XP_011530068.1 41 6912
XM_017007906.1 XP_016863395.1 43 7129
XM_017007907.1 XP_016863396.1 43 7126
XM_017007908.1 XP_016863397.1 42 7058
XM_017007909.1 XP_016863398.1 28 4754
GenBank mRNA AB023210.2 - 5 643
AF538685.1 - 39 6852
BC150246.1 - 5 664

View Full Product Details