Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012341.2 | NP_036473.2 | 13 | 1370 | |
GenBank mRNA | AF120334.1 | - | 13 | 1322 | |
AF325353.1 | - | 13 | 1299 | ||
AK001548.1 | - | 13 | 1317 | ||
AK001552.1 | - | 13 | 1372 | ||
AK097093.1 | - | 11 | 2178 | ||
AK222861.1 | - | 13 | 1317 | ||
AK295231.1 | - | 12 | 1213 | ||
AK301721.1 | - | 13 | 1485 | ||
AK302219.1 | - | 12 | 1175 | ||
BC033784.1 | - | 13 | 1293 | ||
BC038975.2 | - | 13 | 1331 | 5534986 |