Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015509.3 | NP_056324.2 | 3 | 345 | |
NR_024260.1 | 3 | 345 | |||
GenBank mRNA | AK074858.1 | - | 3 | 302 | |
AK074880.1 | - | 3 | 315 | ||
AK074923.1 | - | 3 | 288 | ||
AK075013.1 | - | 3 | 322 | ||
AK290537.1 | - | 3 | 270 | ||
AK303675.1 | - | 3 | 288 | ||
AK314424.1 | - | 3 | 279 | ||
AL050272.1 | - | 3 | 274 | ||
BC002888.2 | - | 3 | 247 | ||
BC067367.1 | - | 1 | 17 | ||
BC110876.1 | - | 3 | 279 | 5553942 | |
DA496321.1 | - | 3 | 345 |