Gene Transcripts

Gene Symbol
GH2
Entrez Gene ID
2689
Gene Name
growth hormone 2
Gene Aliases
GH-V, GHB2, GHL, GHV, hGH-V
Chromosome Location
Chr. 17: 63880212 - 63881942 on Build GRCh38
UniGene ID
Hs.406754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002059.4 NP_002050.1 3 393
NM_022556.3 NP_072050.1 3 348
NM_022557.3 NP_072051.1 3 393
NM_022558.3 NP_072052.1 3 393
GenBank mRNA AB451332.1 - 2 207
AB451477.1 - 2 207
AF006060.1 - 3 219
AF006061.1 - 2 252
AK312194.1 - 3 315
AU139016.1 - 3 393
BC020760.1 - 3 314 4720475
CR541902.1 - 2 252
CR541924.1 - 2 252
J03756.1 - 3 313
M38451.1 - 3 264

View Full Product Details