Gene Transcripts

Gene Symbol
SCP2
Entrez Gene ID
6342
Gene Name
sterol carrier protein 2
Gene Aliases
NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX
Chromosome Location
Chr. 1: 52927229 - 53051617 on Build GRCh38
UniGene ID
Hs.476365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193599.1 NP_001180528.1 3 389
NM_001193617.1 NP_001180546.1 3 403
NM_002979.4 NP_002970.2 4 461
XM_005271103.4 XP_005271160.1 4 404
XM_011541935.2 XP_011540237.1 4 404
XM_017002045.1 XP_016857534.1 4 404
GenBank mRNA AB208789.1 - 4 413
AK294631.1 - 3 332
AK295214.1 - 3 346
AK312856.1 - 4 412
DC330171.1 - 4 445
M75883.1 - 4 314

View Full Product Details