Gene Transcripts

Gene Symbol
ZNF768
Entrez Gene ID
79724
Gene Name
zinc finger protein 768
Gene Aliases
Chromosome Location
Chr. 16: 30524001 - 30532367 on Build GRCh38
UniGene ID
Hs.85658
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024671.3 NP_078947.3 2 471
XM_017023665.1 XP_016879154.1 2 1476
XM_017023666.1 XP_016879155.1 2 296
GenBank mRNA AK027089.1 - 2 243
AK295274.1 - 2 240
AK315449.1 - 2 267
BC013760.1 - 2 471 3856266
BC092403.1 - 2 383 6201079

View Full Product Details