Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020461.3 | NP_065194.2 | 1 | 716 | |
GenBank mRNA | AB051456.1 | - | 1 | 110 | |
AF272887.1 | - | 1 | 309 | ||
AK310419.1 | - | 1 | 750 | ||
AL832617.2 | - | 1 | 737 | ||
BC144488.1 | - | 1 | 308 | 9053015 | |
BC144489.1 | - | 1 | 308 | ||
BC150613.1 | - | 1 | 308 | 9056983 | |
BC151209.1 | - | 1 | 698 | 40147276 |