Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128931.1 | NP_001122403.1 | 5 | 980 | |
NM_138386.2 | NP_612395.2 | 5 | 980 | ||
XM_011532410.2 | XP_011530712.1 | 5 | 4447 | ||
XM_011532411.2 | XP_011530713.1 | 6 | 1054 | ||
GenBank mRNA | AK097352.1 | - | 5 | 890 | |
AK308601.1 | - | 5 | 949 | ||
BC008207.1 | - | 5 | 980 | 3447276 |