Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49061406-49061914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTF4
Entrez Gene ID
4909
Gene Name
neurotrophin 4
Gene Aliases
GLC10, GLC1O, NT-4, NT-4/5, NT-5, NT4, NT5, NTF5
UniGene
Hs.266902
Gene Chromosome Location
Chr.19: 49058284-49065076 on Build GRCh38
Transcripts
NM_006179.4 XM_005258962.3 XM_006723232.3 XM_011527008.2 XM_011527009.2 XM_011527010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403508 c.565G>T p.G189C Chr.19: 49061433-49061433 on Build GRCh38
6402737 c.221C>A p.P74Q Chr.19: 49061777-49061777 on Build GRCh38
1286708 c.474C>T p.C158C Chr.19: 49061524-49061524 on Build GRCh38
6289155 c.328G>A p.A110T Chr.19: 49061670-49061670 on Build GRCh38
2752019 c.452C>T p.P151L Chr.19: 49061546-49061546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951341070 Chr.19: 49061701-49061701 on Build GRCh38
rs775118459 Chr.19: 49061688-49061688 on Build GRCh38
rs556785762 Chr.19: 49061707-49061707 on Build GRCh38
rs774223674 Chr.19: 49061651-49061651 on Build GRCh38
rs771342513 Chr.19: 49061772-49061772 on Build GRCh38
rs776177618 Chr.19: 49061482-49061482 on Build GRCh38
rs748961545 Chr.19: 49061681-49061681 on Build GRCh38
rs1029494815 Chr.19: 49061869-49061869 on Build GRCh38
rs767514523 Chr.19: 49061776-49061776 on Build GRCh38
rs759474455 Chr.19: 49061656-49061656 on Build GRCh38
rs749392406 Chr.19: 49061438-49061438 on Build GRCh38
rs940302777 Chr.19: 49061864-49061864 on Build GRCh38
rs746321607 Chr.19: 49061538-49061538 on Build GRCh38
rs778141660 Chr.19: 49061740-49061740 on Build GRCh38
rs768336179 Chr.19: 49061546-49061546 on Build GRCh38
rs367858798 Chr.19: 49061738-49061738 on Build GRCh38
rs772979004 Chr.19: 49061503-49061503 on Build GRCh38
rs752041542 Chr.19: 49061675-49061675 on Build GRCh38
rs778409311 Chr.19: 49061685-49061685 on Build GRCh38
rs186082918 Chr.19: 49061775-49061775 on Build GRCh38
rs963958335 Chr.19: 49061842-49061842 on Build GRCh38
rs759376040 Chr.19: 49061587-49061587 on Build GRCh38
rs746640305 Chr.19: 49061514-49061514 on Build GRCh38
rs755261332 Chr.19: 49061626-49061626 on Build GRCh38
rs758946318 Chr.19: 49061617-49061617 on Build GRCh38
rs371058489 Chr.19: 49061667-49061667 on Build GRCh38
rs200786116 Chr.19: 49061795-49061795 on Build GRCh38
rs957177310 Chr.19: 49061794-49061794 on Build GRCh38
rs775964511 Chr.19: 49061465-49061466 on Build GRCh38
rs761282348 Chr.19: 49061484-49061484 on Build GRCh38
rs62127897 Chr.19: 49061581-49061581 on Build GRCh38
rs773531060 Chr.19: 49061712-49061712 on Build GRCh38
rs779054536 Chr.19: 49061537-49061537 on Build GRCh38
rs761090241 Chr.19: 49061781-49061781 on Build GRCh38
rs1017274820 Chr.19: 49061841-49061841 on Build GRCh38
rs922351065 Chr.19: 49061519-49061519 on Build GRCh38
rs370717457 Chr.19: 49061466-49061466 on Build GRCh38
rs755690314 Chr.19: 49061679-49061679 on Build GRCh38
rs777449663 Chr.19: 49061680-49061680 on Build GRCh38
rs147294437 Chr.19: 49061446-49061446 on Build GRCh38
rs955141551 Chr.19: 49061870-49061870 on Build GRCh38
rs879094726 Chr.19: 49061715-49061715 on Build GRCh38
rs781620265 Chr.19: 49061631-49061631 on Build GRCh38
rs367957078 Chr.19: 49061481-49061481 on Build GRCh38
rs576716713 Chr.19: 49061852-49061852 on Build GRCh38
rs977260366 Chr.19: 49061600-49061600 on Build GRCh38
rs371019044 Chr.19: 49061721-49061721 on Build GRCh38
rs374367338 Chr.19: 49061732-49061732 on Build GRCh38
rs921018899 Chr.19: 49061796-49061796 on Build GRCh38
rs201069064 Chr.19: 49061580-49061580 on Build GRCh38
rs770833595 Chr.19: 49061649-49061649 on Build GRCh38
rs369484075 Chr.19: 49061619-49061619 on Build GRCh38
rs975549134 Chr.19: 49061515-49061515 on Build GRCh38
rs754267965 Chr.19: 49061872-49061872 on Build GRCh38
rs772790339 Chr.19: 49061571-49061571 on Build GRCh38
rs987875217 Chr.19: 49061874-49061874 on Build GRCh38
rs932386085 Chr.19: 49061805-49061805 on Build GRCh38
rs572767613 Chr.19: 49061749-49061749 on Build GRCh38
rs746418406 Chr.19: 49061453-49061453 on Build GRCh38
rs1033709780 Chr.19: 49061476-49061476 on Build GRCh38
rs762861349 Chr.19: 49061579-49061579 on Build GRCh38
rs974105707 Chr.19: 49061455-49061455 on Build GRCh38
rs1051216517 Chr.19: 49061845-49061845 on Build GRCh38
rs373941167 Chr.19: 49061632-49061632 on Build GRCh38
rs34922845 Chr.19: 49061579-49061580 on Build GRCh38
rs776012757 Chr.19: 49061469-49061469 on Build GRCh38
rs377741315 Chr.19: 49061678-49061678 on Build GRCh38
rs200878406 Chr.19: 49061499-49061499 on Build GRCh38
rs887767311 Chr.19: 49061783-49061783 on Build GRCh38
rs759034173 Chr.19: 49061540-49061540 on Build GRCh38
rs374345134 Chr.19: 49061545-49061545 on Build GRCh38
rs772499451 Chr.19: 49061457-49061457 on Build GRCh38
rs369066724 Chr.19: 49061517-49061517 on Build GRCh38
rs912207320 Chr.19: 49061846-49061846 on Build GRCh38
rs142027939 Chr.19: 49061437-49061437 on Build GRCh38
rs202216349 Chr.19: 49061687-49061687 on Build GRCh38
rs374113081 Chr.19: 49061671-49061671 on Build GRCh38
rs797044939 Chr.19: 49061477-49061477 on Build GRCh38
rs770388056 Chr.19: 49061684-49061684 on Build GRCh38
rs552406043 Chr.19: 49061614-49061614 on Build GRCh38
rs764829441 Chr.19: 49061486-49061486 on Build GRCh38
rs766504681 Chr.19: 49061729-49061729 on Build GRCh38
rs755518603 Chr.19: 49061676-49061676 on Build GRCh38
rs375526828 Chr.19: 49061433-49061433 on Build GRCh38
rs766101589 Chr.19: 49061509-49061509 on Build GRCh38
rs763358860 Chr.19: 49061493-49061494 on Build GRCh38
rs1037232374 Chr.19: 49061878-49061878 on Build GRCh38
rs780565192 Chr.19: 49061541-49061541 on Build GRCh38
rs768503189 Chr.19: 49061762-49061762 on Build GRCh38
rs748834740 Chr.19: 49061646-49061646 on Build GRCh38
rs749505090 Chr.19: 49061769-49061769 on Build GRCh38
rs924409835 Chr.19: 49061608-49061608 on Build GRCh38
rs913593395 Chr.19: 49061888-49061888 on Build GRCh38
rs756962734 Chr.19: 49061748-49061748 on Build GRCh38
rs371734511 Chr.19: 49061505-49061505 on Build GRCh38
rs898702012 Chr.19: 49061890-49061890 on Build GRCh38
rs776024123 Chr.19: 49061779-49061779 on Build GRCh38
rs753431367 Chr.19: 49061745-49061745 on Build GRCh38
rs757515516 Chr.19: 49061607-49061607 on Build GRCh38
rs570895408 Chr.19: 49061637-49061637 on Build GRCh38
rs865923726 Chr.19: 49061500-49061500 on Build GRCh38
rs554407199 Chr.19: 49061742-49061742 on Build GRCh38
rs61732310 Chr.19: 49061735-49061735 on Build GRCh38
rs751743400 Chr.19: 49061730-49061730 on Build GRCh38
rs774261204 Chr.19: 49061774-49061774 on Build GRCh38
rs764831671 Chr.19: 49061493-49061493 on Build GRCh38
rs771706948 Chr.19: 49061657-49061657 on Build GRCh38
rs145884553 Chr.19: 49061755-49061755 on Build GRCh38
rs776576559 Chr.19: 49061768-49061768 on Build GRCh38
rs769115282 Chr.19: 49061563-49061563 on Build GRCh38
rs144842220 Chr.19: 49061601-49061601 on Build GRCh38
rs1004394151 Chr.19: 49061726-49061726 on Build GRCh38
rs747545895 Chr.19: 49061622-49061622 on Build GRCh38
rs374415552 Chr.19: 49061471-49061471 on Build GRCh38
rs750511722 Chr.19: 49061670-49061670 on Build GRCh38
rs764263332 Chr.19: 49061523-49061523 on Build GRCh38
rs140946623 Chr.19: 49061562-49061562 on Build GRCh38
rs753931867 Chr.19: 49061527-49061527 on Build GRCh38
rs145846156 Chr.19: 49061618-49061618 on Build GRCh38
rs924460891 Chr.19: 49061777-49061777 on Build GRCh38
rs377553005 Chr.19: 49061658-49061658 on Build GRCh38
rs746954793 Chr.19: 49061760-49061760 on Build GRCh38
rs750392853 Chr.19: 49061655-49061655 on Build GRCh38
rs1015733961 Chr.19: 49061727-49061727 on Build GRCh38
rs757244498 Chr.19: 49061535-49061535 on Build GRCh38
rs751306303 Chr.19: 49061734-49061734 on Build GRCh38
rs767678419 Chr.19: 49061750-49061750 on Build GRCh38
rs779336456 Chr.19: 49061441-49061441 on Build GRCh38
rs767366652 Chr.19: 49061589-49061589 on Build GRCh38
rs752948219 Chr.19: 49061756-49061756 on Build GRCh38
rs144346396 Chr.19: 49061439-49061439 on Build GRCh38
rs199958178 Chr.19: 49061704-49061704 on Build GRCh38
rs769032965 Chr.19: 49061726-49061727 on Build GRCh38
rs933661685 Chr.19: 49061531-49061531 on Build GRCh38

View Full Product Details