Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97733187-97733676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
UniGene
Hs.234569
Gene Chromosome Location
Chr.2: 97712030-97744327 on Build GRCh38
Transcripts
NM_001079.3 XM_017004867.1 XM_017004868.1 XM_017004869.1 XM_017004870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6267695 c.790+7G>T p.? Chr.2: 97733219-97733219 on Build GRCh38
5736138 c.847C>T p.R283* Chr.2: 97733553-97733553 on Build GRCh38
6019816 c.788C>T p.S263L Chr.2: 97733210-97733210 on Build GRCh38
1233086 c.853G>A p.D285N Chr.2: 97733559-97733559 on Build GRCh38
6092725 c.450G>T p.T150T Chr.2: 97733334-97733334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553261328 Chr.2: 97733558-97733558 on Build GRCh38
rs200585997 Chr.2: 97733512-97733512 on Build GRCh38
rs759650270 Chr.2: 97733343-97733343 on Build GRCh38
rs761700835 Chr.2: 97733520-97733520 on Build GRCh38
rs757286783 Chr.2: 97733291-97733291 on Build GRCh38
rs201810987 Chr.2: 97733617-97733617 on Build GRCh38
rs761189828 Chr.2: 97733260-97733260 on Build GRCh38
rs756672820 Chr.2: 97733585-97733585 on Build GRCh38
rs143181735 Chr.2: 97733316-97733316 on Build GRCh38
rs373266022 Chr.2: 97733227-97733227 on Build GRCh38
rs774406382 Chr.2: 97733508-97733508 on Build GRCh38
rs760529841 Chr.2: 97733614-97733614 on Build GRCh38
rs200018109 Chr.2: 97733218-97733218 on Build GRCh38
rs750129763 Chr.2: 97733438-97733440 on Build GRCh38
rs780517718 Chr.2: 97733589-97733589 on Build GRCh38
rs753453013 Chr.2: 97733548-97733548 on Build GRCh38
rs200126170 Chr.2: 97733572-97733572 on Build GRCh38
rs924316720 Chr.2: 97733387-97733387 on Build GRCh38
rs111487075 Chr.2: 97733447-97733447 on Build GRCh38
rs765489716 Chr.2: 97733276-97733276 on Build GRCh38
rs201776628 Chr.2: 97733341-97733341 on Build GRCh38
rs55715517 Chr.2: 97733437-97733437 on Build GRCh38
rs762766552 Chr.2: 97733280-97733280 on Build GRCh38
rs557179479 Chr.2: 97733506-97733506 on Build GRCh38
rs201017686 Chr.2: 97733609-97733609 on Build GRCh38
rs759115124 Chr.2: 97733619-97733619 on Build GRCh38
rs112357205 Chr.2: 97733493-97733493 on Build GRCh38
rs1017869021 Chr.2: 97733281-97733281 on Build GRCh38
rs982143659 Chr.2: 97733480-97733480 on Build GRCh38
rs754023614 Chr.2: 97733226-97733226 on Build GRCh38
rs764218522 Chr.2: 97733375-97733375 on Build GRCh38
rs1008189958 Chr.2: 97733653-97733653 on Build GRCh38
rs779657217 Chr.2: 97733432-97733432 on Build GRCh38
rs769875156 Chr.2: 97733486-97733486 on Build GRCh38
rs55711283 Chr.2: 97733229-97733229 on Build GRCh38
rs55852368 Chr.2: 97733228-97733228 on Build GRCh38
rs771165725 Chr.2: 97733474-97733474 on Build GRCh38
rs544647865 Chr.2: 97733615-97733615 on Build GRCh38
rs140012719 Chr.2: 97733328-97733328 on Build GRCh38
rs201761684 Chr.2: 97733232-97733232 on Build GRCh38
rs767532399 Chr.2: 97733407-97733407 on Build GRCh38
rs766370265 Chr.2: 97733262-97733262 on Build GRCh38
rs201414967 Chr.2: 97733285-97733285 on Build GRCh38
rs764977425 Chr.2: 97733554-97733554 on Build GRCh38
rs750348430 Chr.2: 97733423-97733423 on Build GRCh38
rs370478398 Chr.2: 97733498-97733498 on Build GRCh38
rs113994173 Chr.2: 97733464-97733464 on Build GRCh38
rs781477800 Chr.2: 97733559-97733559 on Build GRCh38
rs370811702 Chr.2: 97733607-97733607 on Build GRCh38
rs749776847 Chr.2: 97733268-97733268 on Build GRCh38
rs374930707 Chr.2: 97733399-97733399 on Build GRCh38
rs201824457 Chr.2: 97733620-97733620 on Build GRCh38
rs778413836 Chr.2: 97733449-97733449 on Build GRCh38
rs755711247 Chr.2: 97733429-97733429 on Build GRCh38
rs56133341 Chr.2: 97733217-97733217 on Build GRCh38
rs916598301 Chr.2: 97733208-97733208 on Build GRCh38
rs755656119 Chr.2: 97733295-97733295 on Build GRCh38
rs748903250 Chr.2: 97733310-97733310 on Build GRCh38
rs200666396 Chr.2: 97733404-97733404 on Build GRCh38
rs764893160 Chr.2: 97733402-97733413 on Build GRCh38
rs199639189 Chr.2: 97733349-97733349 on Build GRCh38
rs199502727 Chr.2: 97733333-97733333 on Build GRCh38
rs202121787 Chr.2: 97733231-97733231 on Build GRCh38
rs909677452 Chr.2: 97733584-97733584 on Build GRCh38
rs772340901 Chr.2: 97733313-97733313 on Build GRCh38
rs776834138 Chr.2: 97733269-97733269 on Build GRCh38
rs200657253 Chr.2: 97733356-97733356 on Build GRCh38
rs202079520 Chr.2: 97733505-97733505 on Build GRCh38
rs775710963 Chr.2: 97733360-97733360 on Build GRCh38
rs993494098 Chr.2: 97733633-97733633 on Build GRCh38
rs762482538 Chr.2: 97733445-97733450 on Build GRCh38
rs781685153 Chr.2: 97733475-97733475 on Build GRCh38
rs753549375 Chr.2: 97733436-97733436 on Build GRCh38
rs572701573 Chr.2: 97733212-97733212 on Build GRCh38
rs751464383 Chr.2: 97733286-97733286 on Build GRCh38
rs546937496 Chr.2: 97733243-97733243 on Build GRCh38
rs200437787 Chr.2: 97733626-97733626 on Build GRCh38
rs759596877 Chr.2: 97733272-97733272 on Build GRCh38
rs991422899 Chr.2: 97733352-97733352 on Build GRCh38
rs374670472 Chr.2: 97733382-97733382 on Build GRCh38
rs763176121 Chr.2: 97733370-97733370 on Build GRCh38
rs200251859 Chr.2: 97733235-97733235 on Build GRCh38
rs200358194 Chr.2: 97733230-97733230 on Build GRCh38
rs768188594 Chr.2: 97733206-97733206 on Build GRCh38
rs760626189 Chr.2: 97733544-97733544 on Build GRCh38
rs200987919 Chr.2: 97733632-97733632 on Build GRCh38
rs755100834 Chr.2: 97733647-97733647 on Build GRCh38
rs780571160 Chr.2: 97733643-97733643 on Build GRCh38
rs774658245 Chr.2: 97733603-97733603 on Build GRCh38
rs536451792 Chr.2: 97733527-97733527 on Build GRCh38
rs746378071 Chr.2: 97733579-97733579 on Build GRCh38
rs761349565 Chr.2: 97733221-97733222 on Build GRCh38
rs747446900 Chr.2: 97733459-97733459 on Build GRCh38
rs188148020 Chr.2: 97733642-97733642 on Build GRCh38
rs773298062 Chr.2: 97733539-97733539 on Build GRCh38
rs201457490 Chr.2: 97733601-97733601 on Build GRCh38
rs749711251 Chr.2: 97733646-97733646 on Build GRCh38
rs199710795 Chr.2: 97733242-97733242 on Build GRCh38
rs56096305 Chr.2: 97733602-97733602 on Build GRCh38
rs746322954 Chr.2: 97733482-97733482 on Build GRCh38
rs200430850 Chr.2: 97733294-97733294 on Build GRCh38
rs377720729 Chr.2: 97733247-97733247 on Build GRCh38
rs200501678 Chr.2: 97733641-97733641 on Build GRCh38
rs772445297 Chr.2: 97733249-97733249 on Build GRCh38
rs955695637 Chr.2: 97733391-97733391 on Build GRCh38
rs754420169 Chr.2: 97733553-97733553 on Build GRCh38
rs773211731 Chr.2: 97733610-97733610 on Build GRCh38
rs773619110 Chr.2: 97733251-97733251 on Build GRCh38
rs56033246 Chr.2: 97733236-97733236 on Build GRCh38
rs779887992 Chr.2: 97733299-97733299 on Build GRCh38
rs955067565 Chr.2: 97733329-97733329 on Build GRCh38
rs56081234 Chr.2: 97733638-97733638 on Build GRCh38
rs200797733 Chr.2: 97733334-97733334 on Build GRCh38
rs201225206 Chr.2: 97733592-97733592 on Build GRCh38
rs897858343 Chr.2: 97733618-97733618 on Build GRCh38
rs780978714 Chr.2: 97733293-97733293 on Build GRCh38
rs897827672 Chr.2: 97733270-97733270 on Build GRCh38
rs117994264 Chr.2: 97733398-97733398 on Build GRCh38

View Full Product Details