Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97737389-97737890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
UniGene
Hs.234569
Gene Chromosome Location
Chr.2: 97712030-97744327 on Build GRCh38
Transcripts
NM_001079.3 NM_207519.1 XM_017004867.1 XM_017004868.1 XM_017004869.1 XM_017004870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3669026 c.1417C>G p.H473D Chr.2: 97737600-97737600 on Build GRCh38
4883775 c.1404G>T p.L468L Chr.2: 97737587-97737587 on Build GRCh38
6542851 c.1014C>T p.A338A Chr.2: 97737575-97737575 on Build GRCh38
6542850 c.1392C>T p.A464A Chr.2: 97737575-97737575 on Build GRCh38
4187094 c.1348A>C p.K450Q Chr.2: 97737531-97737531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780586765 Chr.2: 97737823-97737823 on Build GRCh38
rs893210309 Chr.2: 97737422-97737422 on Build GRCh38
rs758807149 Chr.2: 97737598-97737598 on Build GRCh38
rs747461254 Chr.2: 97737490-97737490 on Build GRCh38
rs200149598 Chr.2: 97737726-97737726 on Build GRCh38
rs1056183383 Chr.2: 97737521-97737521 on Build GRCh38
rs201580376 Chr.2: 97737659-97737659 on Build GRCh38
rs959265564 Chr.2: 97737829-97737829 on Build GRCh38
rs749528047 Chr.2: 97737729-97737729 on Build GRCh38
rs771007414 Chr.2: 97737500-97737500 on Build GRCh38
rs766082644 Chr.2: 97737582-97737582 on Build GRCh38
rs201830903 Chr.2: 97737709-97737709 on Build GRCh38
rs770902102 Chr.2: 97737760-97737760 on Build GRCh38
rs55706605 Chr.2: 97737690-97737690 on Build GRCh38
rs770985198 Chr.2: 97737800-97737801 on Build GRCh38
rs756841049 Chr.2: 97737435-97737435 on Build GRCh38
rs201936638 Chr.2: 97737617-97737617 on Build GRCh38
rs779242940 Chr.2: 97737736-97737736 on Build GRCh38
rs201266526 Chr.2: 97737597-97737597 on Build GRCh38
rs104893674 Chr.2: 97737828-97737828 on Build GRCh38
rs140077233 Chr.2: 97737665-97737665 on Build GRCh38
rs760467073 Chr.2: 97737753-97737753 on Build GRCh38
rs374216712 Chr.2: 97737455-97737455 on Build GRCh38
rs367624923 Chr.2: 97737611-97737611 on Build GRCh38
rs17488834 Chr.2: 97737676-97737676 on Build GRCh38
rs199994054 Chr.2: 97737606-97737606 on Build GRCh38
rs113994174 Chr.2: 97737576-97737576 on Build GRCh38
rs199539961 Chr.2: 97737691-97737691 on Build GRCh38
rs746810754 Chr.2: 97737864-97737864 on Build GRCh38
rs764911782 Chr.2: 97737768-97737768 on Build GRCh38
rs369459242 Chr.2: 97737702-97737702 on Build GRCh38
rs149765220 Chr.2: 97737764-97737764 on Build GRCh38
rs762187360 Chr.2: 97737786-97737786 on Build GRCh38
rs767067866 Chr.2: 97737453-97737453 on Build GRCh38
rs946895299 Chr.2: 97737433-97737433 on Build GRCh38
rs751972851 Chr.2: 97737694-97737694 on Build GRCh38
rs751413139 Chr.2: 97737427-97737427 on Build GRCh38
rs55997284 Chr.2: 97737650-97737650 on Build GRCh38
rs781688152 Chr.2: 97737707-97737707 on Build GRCh38
rs767813141 Chr.2: 97737789-97737789 on Build GRCh38
rs201082245 Chr.2: 97737729-97737729 on Build GRCh38
rs150631046 Chr.2: 97737761-97737761 on Build GRCh38
rs775805179 Chr.2: 97737637-97737637 on Build GRCh38
rs137853201 Chr.2: 97737577-97737577 on Build GRCh38
rs867742833 Chr.2: 97737778-97737778 on Build GRCh38
rs771994200 Chr.2: 97737841-97737841 on Build GRCh38
rs201003454 Chr.2: 97737830-97737830 on Build GRCh38
rs1054368473 Chr.2: 97737680-97737680 on Build GRCh38
rs774434467 Chr.2: 97737571-97737571 on Build GRCh38
rs184903382 Chr.2: 97737834-97737834 on Build GRCh38
rs56059280 Chr.2: 97737651-97737651 on Build GRCh38
rs750880509 Chr.2: 97737710-97737710 on Build GRCh38
rs749947670 Chr.2: 97737454-97737454 on Build GRCh38
rs200712258 Chr.2: 97737463-97737463 on Build GRCh38
rs745937123 Chr.2: 97737507-97737507 on Build GRCh38
rs765744939 Chr.2: 97737686-97737686 on Build GRCh38
rs778361599 Chr.2: 97737488-97737488 on Build GRCh38
rs200439854 Chr.2: 97737688-97737688 on Build GRCh38
rs777987151 Chr.2: 97737472-97737473 on Build GRCh38
rs200348949 Chr.2: 97737647-97737647 on Build GRCh38
rs201906752 Chr.2: 97737501-97737501 on Build GRCh38
rs200787273 Chr.2: 97737693-97737693 on Build GRCh38
rs56146954 Chr.2: 97737854-97737854 on Build GRCh38
rs199873761 Chr.2: 97737581-97737581 on Build GRCh38
rs773242018 Chr.2: 97737672-97737672 on Build GRCh38
rs201814622 Chr.2: 97737462-97737462 on Build GRCh38
rs757588248 Chr.2: 97737698-97737698 on Build GRCh38
rs777782165 Chr.2: 97737851-97737851 on Build GRCh38
rs770105318 Chr.2: 97737508-97737508 on Build GRCh38
rs201117726 Chr.2: 97737615-97737615 on Build GRCh38
rs747985410 Chr.2: 97737748-97737748 on Build GRCh38
rs775546124 Chr.2: 97737522-97737522 on Build GRCh38
rs749373144 Chr.2: 97737718-97737718 on Build GRCh38
rs201902235 Chr.2: 97737725-97737725 on Build GRCh38
rs373787406 Chr.2: 97737516-97737516 on Build GRCh38
rs201016309 Chr.2: 97737783-97737783 on Build GRCh38
rs200758703 Chr.2: 97737715-97737715 on Build GRCh38
rs199639448 Chr.2: 97737713-97737713 on Build GRCh38
rs771940938 Chr.2: 97737749-97737749 on Build GRCh38
rs202012936 Chr.2: 97737605-97737605 on Build GRCh38
rs763747511 Chr.2: 97737569-97737569 on Build GRCh38
rs199775081 Chr.2: 97737504-97737504 on Build GRCh38
rs750794851 Chr.2: 97737792-97737792 on Build GRCh38
rs201451319 Chr.2: 97737737-97737737 on Build GRCh38
rs730880319 Chr.2: 97737784-97737796 on Build GRCh38
rs752308469 Chr.2: 97737780-97737780 on Build GRCh38
rs756232467 Chr.2: 97737714-97737714 on Build GRCh38
rs779549199 Chr.2: 97737457-97737457 on Build GRCh38
rs200693152 Chr.2: 97737536-97737536 on Build GRCh38
rs915506858 Chr.2: 97737727-97737727 on Build GRCh38
rs550812644 Chr.2: 97737799-97737799 on Build GRCh38
rs748318780 Chr.2: 97737835-97737835 on Build GRCh38
rs371850086 Chr.2: 97737626-97737626 on Build GRCh38
rs200914614 Chr.2: 97737649-97737649 on Build GRCh38
rs777825759 Chr.2: 97737747-97737747 on Build GRCh38
rs542619486 Chr.2: 97737482-97737482 on Build GRCh38
rs55969019 Chr.2: 97737725-97737725 on Build GRCh38
rs752820490 Chr.2: 97737825-97737825 on Build GRCh38
rs56189815 Chr.2: 97737842-97737842 on Build GRCh38
rs1012199340 Chr.2: 97737563-97737563 on Build GRCh38
rs201712942 Chr.2: 97737572-97737572 on Build GRCh38

View Full Product Details