Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154551747-154552266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKBKG
Entrez Gene ID
8517
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
Gene Aliases
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9
UniGene
Hs.43505
Gene Chromosome Location
Chr.X: 154542240-154565046 on Build GRCh38
Transcripts
NM_001099856.4 NM_001099857.2 NM_001145255.2 NM_001321396.1 NM_001321397.1 NM_003639.4

SNP Detail

SNP ID Chromosome Location
rs901820688 Chr.X: 154552165-154552165 on Build GRCh38
rs376019792 Chr.X: 154552008-154552008 on Build GRCh38
rs782603646 Chr.X: 154551915-154551915 on Build GRCh38
rs991143065 Chr.X: 154551954-154551954 on Build GRCh38
rs782515018 Chr.X: 154551875-154551875 on Build GRCh38
rs782533747 Chr.X: 154552058-154552058 on Build GRCh38
rs149511938 Chr.X: 154552140-154552140 on Build GRCh38
rs966867546 Chr.X: 154551907-154551907 on Build GRCh38
rs782576988 Chr.X: 154552025-154552025 on Build GRCh38
rs1001035466 Chr.X: 154552243-154552243 on Build GRCh38
rs782276373 Chr.X: 154551939-154551939 on Build GRCh38
rs782455868 Chr.X: 154552086-154552086 on Build GRCh38
rs782480682 Chr.X: 154551868-154551868 on Build GRCh38
rs782039548 Chr.X: 154551981-154551981 on Build GRCh38
rs200122243 Chr.X: 154551876-154551876 on Build GRCh38
rs782141440 Chr.X: 154552135-154552135 on Build GRCh38
rs782717047 Chr.X: 154552151-154552151 on Build GRCh38
rs782167422 Chr.X: 154552122-154552122 on Build GRCh38
rs782803636 Chr.X: 154551946-154551946 on Build GRCh38
rs377406996 Chr.X: 154552153-154552153 on Build GRCh38
rs57174646 Chr.X: 154552009-154552009 on Build GRCh38
rs782663126 Chr.X: 154552062-154552062 on Build GRCh38
rs782816286 Chr.X: 154551985-154551985 on Build GRCh38
rs966246135 Chr.X: 154551788-154551788 on Build GRCh38
rs371203594 Chr.X: 154551969-154551969 on Build GRCh38
rs781881888 Chr.X: 154551988-154551988 on Build GRCh38
rs782604431 Chr.X: 154552187-154552187 on Build GRCh38
rs782098796 Chr.X: 154551963-154551963 on Build GRCh38
rs781926607 Chr.X: 154551979-154551979 on Build GRCh38
rs782087958 Chr.X: 154552145-154552145 on Build GRCh38
rs137853323 Chr.X: 154552186-154552186 on Build GRCh38
rs1039026297 Chr.X: 154552159-154552159 on Build GRCh38
rs782030098 Chr.X: 154552123-154552123 on Build GRCh38
rs148695964 Chr.X: 154552171-154552171 on Build GRCh38
rs782449420 Chr.X: 154552232-154552232 on Build GRCh38
rs782454227 Chr.X: 154552015-154552015 on Build GRCh38
rs781827676 Chr.X: 154552011-154552011 on Build GRCh38
rs782297181 Chr.X: 154552048-154552048 on Build GRCh38
rs782517338 Chr.X: 154552175-154552175 on Build GRCh38
rs1032761701 Chr.X: 154551945-154551945 on Build GRCh38
rs782594565 Chr.X: 154552085-154552085 on Build GRCh38
rs902721291 Chr.X: 154552206-154552206 on Build GRCh38
rs781985461 Chr.X: 154551961-154551961 on Build GRCh38
rs782355825 Chr.X: 154552077-154552077 on Build GRCh38
rs143999678 Chr.X: 154552141-154552141 on Build GRCh38
rs782236286 Chr.X: 154552064-154552064 on Build GRCh38
rs782813189 Chr.X: 154552163-154552163 on Build GRCh38
rs919093186 Chr.X: 154552061-154552061 on Build GRCh38
rs782173858 Chr.X: 154552160-154552160 on Build GRCh38
rs182403705 Chr.X: 154551790-154551790 on Build GRCh38
rs782221780 Chr.X: 154552210-154552210 on Build GRCh38

View Full Product Details