Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32443357-32443897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRA
Entrez Gene ID
3122
Gene Name
major histocompatibility complex, class II, DR alpha
Gene Aliases
HLA-DRA1, MLRW
UniGene
Hs.520048
Gene Chromosome Location
Chr.6: 32439842-32445046 on Build GRCh38
Transcripts
NM_019111.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5034994 c.512G>A p.R171H Chr.6: 32443368-32443368 on Build GRCh38
5512233 c.613_614delTT p.F205fs*1 Chr.6: 32443758-32443759 on Build GRCh38
4901056 c.580G>A p.G194S Chr.6: 32443436-32443436 on Build GRCh38
224924 c.632T>C p.L211P Chr.6: 32443777-32443777 on Build GRCh38
4997786 c.568G>T p.V190L Chr.6: 32443424-32443424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755315159 Chr.6: 32443831-32443831 on Build GRCh38
rs879138868 Chr.6: 32443469-32443469 on Build GRCh38
rs13218331 Chr.6: 32443585-32443585 on Build GRCh38
rs960870983 Chr.6: 32443871-32443871 on Build GRCh38
rs879185925 Chr.6: 32443754-32443754 on Build GRCh38
rs146370028 Chr.6: 32443388-32443388 on Build GRCh38
rs777243715 Chr.6: 32443848-32443848 on Build GRCh38
rs917732578 Chr.6: 32443659-32443659 on Build GRCh38
rs11544315 Chr.6: 32443796-32443796 on Build GRCh38
rs201633626 Chr.6: 32443687-32443687 on Build GRCh38
rs200798853 Chr.6: 32443409-32443409 on Build GRCh38
rs139874048 Chr.6: 32443808-32443808 on Build GRCh38
rs752606047 Chr.6: 32443414-32443414 on Build GRCh38
rs548267071 Chr.6: 32443550-32443550 on Build GRCh38
rs916798925 Chr.6: 32443657-32443657 on Build GRCh38
rs375630609 Chr.6: 32443704-32443704 on Build GRCh38
rs371441506 Chr.6: 32443512-32443512 on Build GRCh38
rs760381879 Chr.6: 32443381-32443381 on Build GRCh38
rs778808460 Chr.6: 32443675-32443675 on Build GRCh38
rs557232047 Chr.6: 32443583-32443583 on Build GRCh38
rs117531802 Chr.6: 32443613-32443613 on Build GRCh38
rs199723815 Chr.6: 32443433-32443433 on Build GRCh38
rs762948804 Chr.6: 32443872-32443872 on Build GRCh38
rs766104968 Chr.6: 32443742-32443742 on Build GRCh38
rs960905364 Chr.6: 32443602-32443602 on Build GRCh38
rs537246240 Chr.6: 32443470-32443470 on Build GRCh38
rs576887908 Chr.6: 32443691-32443691 on Build GRCh38
rs879014311 Chr.6: 32443537-32443540 on Build GRCh38
rs770380561 Chr.6: 32443853-32443853 on Build GRCh38
rs997782030 Chr.6: 32443392-32443392 on Build GRCh38
rs771561521 Chr.6: 32443511-32443511 on Build GRCh38
rs552360101 Chr.6: 32443472-32443472 on Build GRCh38
rs553117732 Chr.6: 32443523-32443523 on Build GRCh38
rs368601450 Chr.6: 32443778-32443778 on Build GRCh38
rs757405321 Chr.6: 32443615-32443615 on Build GRCh38
rs759121846 Chr.6: 32443867-32443867 on Build GRCh38
rs111492440 Chr.6: 32443707-32443707 on Build GRCh38
rs139677040 Chr.6: 32443749-32443749 on Build GRCh38
rs758466353 Chr.6: 32443759-32443759 on Build GRCh38
rs372692651 Chr.6: 32443431-32443431 on Build GRCh38
rs746125136 Chr.6: 32443733-32443733 on Build GRCh38
rs370989285 Chr.6: 32443735-32443735 on Build GRCh38
rs77288165 Chr.6: 32443753-32443753 on Build GRCh38
rs946733940 Chr.6: 32443666-32443666 on Build GRCh38
rs7192 Chr.6: 32443869-32443869 on Build GRCh38
rs776279402 Chr.6: 32443863-32443863 on Build GRCh38
rs770825671 Chr.6: 32443415-32443415 on Build GRCh38
rs754750691 Chr.6: 32443569-32443569 on Build GRCh38
rs764172368 Chr.6: 32443873-32443873 on Build GRCh38
rs149917607 Chr.6: 32443847-32443847 on Build GRCh38
rs959510329 Chr.6: 32443600-32443600 on Build GRCh38
rs541798926 Chr.6: 32443868-32443868 on Build GRCh38
rs60110771 Chr.6: 32443509-32443509 on Build GRCh38
rs991037547 Chr.6: 32443608-32443608 on Build GRCh38
rs995975632 Chr.6: 32443766-32443766 on Build GRCh38
rs1006122813 Chr.6: 32443799-32443799 on Build GRCh38
rs374588694 Chr.6: 32443740-32443740 on Build GRCh38
rs762199845 Chr.6: 32443747-32443748 on Build GRCh38
rs749480059 Chr.6: 32443826-32443826 on Build GRCh38
rs780271812 Chr.6: 32443819-32443819 on Build GRCh38
rs371727409 Chr.6: 32443748-32443748 on Build GRCh38
rs574757583 Chr.6: 32443528-32443528 on Build GRCh38
rs751562870 Chr.6: 32443875-32443875 on Build GRCh38
rs962124507 Chr.6: 32443531-32443531 on Build GRCh38
rs776171876 Chr.6: 32443744-32443744 on Build GRCh38
rs753708036 Chr.6: 32443408-32443408 on Build GRCh38
rs779207648 Chr.6: 32443838-32443838 on Build GRCh38
rs977938714 Chr.6: 32443728-32443728 on Build GRCh38
rs780246792 Chr.6: 32443451-32443451 on Build GRCh38
rs368192225 Chr.6: 32443732-32443732 on Build GRCh38
rs756467266 Chr.6: 32443447-32443447 on Build GRCh38
rs774535351 Chr.6: 32443478-32443479 on Build GRCh38
rs748653096 Chr.6: 32443843-32443843 on Build GRCh38
rs746475812 Chr.6: 32443852-32443852 on Build GRCh38
rs749707818 Chr.6: 32443461-32443461 on Build GRCh38
rs2239804 Chr.6: 32443746-32443746 on Build GRCh38
rs2239806 Chr.6: 32443530-32443530 on Build GRCh38
rs2239805 Chr.6: 32443599-32443599 on Build GRCh38
rs576697175 Chr.6: 32443561-32443561 on Build GRCh38

View Full Product Details