Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32444926-32445455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRA
Entrez Gene ID
3122
Gene Name
major histocompatibility complex, class II, DR alpha
Gene Aliases
HLA-DRA1, MLRW
UniGene
Hs.520048
Gene Chromosome Location
Chr.6: 32439842-32445046 on Build GRCh38
Transcripts
NM_019111.4

SNP Detail

SNP ID Chromosome Location
rs372671262 Chr.6: 32445252-32445252 on Build GRCh38
rs563502372 Chr.6: 32445283-32445283 on Build GRCh38
rs963558674 Chr.6: 32445079-32445079 on Build GRCh38
rs948351630 Chr.6: 32445169-32445169 on Build GRCh38
rs773693769 Chr.6: 32445100-32445100 on Build GRCh38
rs191184489 Chr.6: 32445096-32445096 on Build GRCh38
rs958984448 Chr.6: 32445129-32445129 on Build GRCh38
rs904256639 Chr.6: 32445198-32445198 on Build GRCh38
rs996497312 Chr.6: 32445000-32445000 on Build GRCh38
rs1014605522 Chr.6: 32445087-32445087 on Build GRCh38
rs977603728 Chr.6: 32445301-32445301 on Build GRCh38
rs3135388 Chr.6: 32445274-32445274 on Build GRCh38
rs73729232 Chr.6: 32445084-32445084 on Build GRCh38
rs890392577 Chr.6: 32445386-32445386 on Build GRCh38
rs867152980 Chr.6: 32445186-32445186 on Build GRCh38
rs1063590 Chr.6: 32444966-32444966 on Build GRCh38
rs147554654 Chr.6: 32445417-32445417 on Build GRCh38
rs1053043 Chr.6: 32445015-32445015 on Build GRCh38
rs1052531 Chr.6: 32444964-32444964 on Build GRCh38
rs992186265 Chr.6: 32445204-32445204 on Build GRCh38
rs1057418434 Chr.6: 32445119-32445119 on Build GRCh38
rs748861155 Chr.6: 32445127-32445127 on Build GRCh38
rs184064011 Chr.6: 32445381-32445381 on Build GRCh38
rs115566509 Chr.6: 32445257-32445257 on Build GRCh38
rs866495193 Chr.6: 32444986-32444986 on Build GRCh38
rs2213585 Chr.6: 32445373-32445373 on Build GRCh38
rs1008585710 Chr.6: 32445047-32445047 on Build GRCh38
rs770394628 Chr.6: 32445202-32445202 on Build GRCh38
rs149878756 Chr.6: 32445352-32445352 on Build GRCh38
rs1050800204 Chr.6: 32445384-32445384 on Build GRCh38
rs1041885 Chr.6: 32445032-32445032 on Build GRCh38
rs377302672 Chr.6: 32445343-32445343 on Build GRCh38
rs62405786 Chr.6: 32445360-32445360 on Build GRCh38
rs1042801409 Chr.6: 32445187-32445187 on Build GRCh38
rs781366068 Chr.6: 32444954-32444954 on Build GRCh38
rs1052916 Chr.6: 32445009-32445009 on Build GRCh38
rs188784615 Chr.6: 32445393-32445393 on Build GRCh38
rs893688624 Chr.6: 32444970-32444970 on Build GRCh38
rs561706764 Chr.6: 32445205-32445205 on Build GRCh38
rs998991915 Chr.6: 32445199-32445199 on Build GRCh38
rs960143488 Chr.6: 32445397-32445397 on Build GRCh38
rs150692352 Chr.6: 32445078-32445078 on Build GRCh38
rs2213586 Chr.6: 32445317-32445317 on Build GRCh38
rs986215091 Chr.6: 32444974-32444974 on Build GRCh38
rs997816417 Chr.6: 32445207-32445207 on Build GRCh38
rs924661757 Chr.6: 32445325-32445325 on Build GRCh38
rs192064884 Chr.6: 32445394-32445394 on Build GRCh38
rs868200881 Chr.6: 32445126-32445126 on Build GRCh38
rs112637929 Chr.6: 32445046-32445046 on Build GRCh38
rs931909310 Chr.6: 32445376-32445376 on Build GRCh38
rs1052644319 Chr.6: 32445201-32445201 on Build GRCh38
rs890318115 Chr.6: 32445035-32445035 on Build GRCh38
rs938642727 Chr.6: 32445097-32445097 on Build GRCh38

View Full Product Details