Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35009479-35010063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL2
Entrez Gene ID
8453
Gene Name
cullin 2
Gene Aliases
-
UniGene
Hs.82919
Gene Chromosome Location
Chr.10: 35008551-35126899 on Build GRCh38
Transcripts
NM_001198777.1 NM_001198778.1 NM_001198779.1 NM_001324375.1 NM_001324376.1 NM_003591.3 XM_011519743.1 XM_011519744.1 XM_011519745.1 XM_011519747.1

SNP Detail

SNP ID Chromosome Location
rs898987789 Chr.10: 35009745-35009745 on Build GRCh38
rs540372788 Chr.10: 35009560-35009560 on Build GRCh38
rs751065740 Chr.10: 35009866-35009866 on Build GRCh38
rs12263386 Chr.10: 35009566-35009566 on Build GRCh38
rs549568727 Chr.10: 35009744-35009744 on Build GRCh38
rs993859421 Chr.10: 35009753-35009753 on Build GRCh38
rs201027415 Chr.10: 35009515-35009515 on Build GRCh38
rs894202312 Chr.10: 35009696-35009696 on Build GRCh38
rs559058619 Chr.10: 35009668-35009668 on Build GRCh38
rs138015093 Chr.10: 35009779-35009779 on Build GRCh38
rs78263878 Chr.10: 35009878-35009878 on Build GRCh38
rs890495510 Chr.10: 35009578-35009578 on Build GRCh38
rs550449231 Chr.10: 35009995-35009995 on Build GRCh38
rs1054346057 Chr.10: 35009992-35009995 on Build GRCh38
rs938344119 Chr.10: 35009679-35009679 on Build GRCh38
rs59269668 Chr.10: 35009513-35009514 on Build GRCh38
rs894069239 Chr.10: 35009542-35009542 on Build GRCh38
rs529522655 Chr.10: 35009690-35009690 on Build GRCh38
rs1026211142 Chr.10: 35009778-35009778 on Build GRCh38
rs942858601 Chr.10: 35009514-35009514 on Build GRCh38
rs947592526 Chr.10: 35009597-35009597 on Build GRCh38
rs763596585 Chr.10: 35009665-35009665 on Build GRCh38
rs1016014190 Chr.10: 35009728-35009728 on Build GRCh38
rs902887968 Chr.10: 35009674-35009674 on Build GRCh38
rs756887696 Chr.10: 35009871-35009871 on Build GRCh38
rs964084871 Chr.10: 35010024-35010024 on Build GRCh38
rs938170881 Chr.10: 35009870-35009870 on Build GRCh38
rs397975596 Chr.10: 35009514-35009515 on Build GRCh38
rs1008984072 Chr.10: 35009640-35009640 on Build GRCh38
rs1012724132 Chr.10: 35009716-35009716 on Build GRCh38
rs982697114 Chr.10: 35009812-35009812 on Build GRCh38
rs963879593 Chr.10: 35009654-35009654 on Build GRCh38
rs755457474 Chr.10: 35009528-35009528 on Build GRCh38
rs975555288 Chr.10: 35009545-35009545 on Build GRCh38
rs15460 Chr.10: 35009984-35009984 on Build GRCh38
rs147061473 Chr.10: 35009992-35009992 on Build GRCh38
rs1020956178 Chr.10: 35009570-35009570 on Build GRCh38
rs558197292 Chr.10: 35009813-35009813 on Build GRCh38
rs986768998 Chr.10: 35009825-35009825 on Build GRCh38
rs1019728546 Chr.10: 35009846-35009846 on Build GRCh38
rs1017705952 Chr.10: 35009647-35009647 on Build GRCh38
rs770982031 Chr.10: 35009869-35009870 on Build GRCh38
rs981791280 Chr.10: 35009842-35009842 on Build GRCh38
rs537152570 Chr.10: 35009827-35009827 on Build GRCh38
rs912636906 Chr.10: 35009510-35009510 on Build GRCh38

View Full Product Details