Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97725051-97725298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
UniGene
Hs.234569
Gene Chromosome Location
Chr.2: 97712030-97744327 on Build GRCh38
Transcripts
NM_001079.3 XM_017004867.1 XM_017004868.1 XM_017004869.1 XM_017004870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
273479 c.413T>C p.L138P Chr.2: 97725102-97725102 on Build GRCh38
7271410 c.405C>T p.G135G Chr.2: 97725094-97725094 on Build GRCh38
2913613 c.431G>A p.S144N Chr.2: 97725120-97725120 on Build GRCh38
7271411 c.27C>T p.G9G Chr.2: 97725094-97725094 on Build GRCh38
7329203 c.34C>A p.L12M Chr.2: 97725101-97725101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61735392 Chr.2: 97725136-97725136 on Build GRCh38
rs770859813 Chr.2: 97725269-97725269 on Build GRCh38
rs760377325 Chr.2: 97725076-97725076 on Build GRCh38
rs776500519 Chr.2: 97725277-97725277 on Build GRCh38
rs199747713 Chr.2: 97725106-97725106 on Build GRCh38
rs773511622 Chr.2: 97725263-97725263 on Build GRCh38
rs779279138 Chr.2: 97725247-97725247 on Build GRCh38
rs201296225 Chr.2: 97725256-97725256 on Build GRCh38
rs56404668 Chr.2: 97725163-97725163 on Build GRCh38
rs766465030 Chr.2: 97725082-97725082 on Build GRCh38
rs752305377 Chr.2: 97725102-97725102 on Build GRCh38
rs886056485 Chr.2: 97725205-97725205 on Build GRCh38
rs55965088 Chr.2: 97725175-97725175 on Build GRCh38
rs199631517 Chr.2: 97725201-97725201 on Build GRCh38
rs201480172 Chr.2: 97725192-97725192 on Build GRCh38
rs746301498 Chr.2: 97725128-97725128 on Build GRCh38
rs56371202 Chr.2: 97725134-97725134 on Build GRCh38
rs115846138 Chr.2: 97725220-97725220 on Build GRCh38
rs749801885 Chr.2: 97725231-97725231 on Build GRCh38
rs368446882 Chr.2: 97725129-97725129 on Build GRCh38
rs758002455 Chr.2: 97725112-97725112 on Build GRCh38
rs780338175 Chr.2: 97725226-97725226 on Build GRCh38
rs372103325 Chr.2: 97725232-97725232 on Build GRCh38
rs921816880 Chr.2: 97725174-97725174 on Build GRCh38
rs772011597 Chr.2: 97725260-97725260 on Build GRCh38
rs778513394 Chr.2: 97725099-97725099 on Build GRCh38
rs756495134 Chr.2: 97725224-97725224 on Build GRCh38
rs753793928 Chr.2: 97725085-97725085 on Build GRCh38
rs202119308 Chr.2: 97725119-97725119 on Build GRCh38
rs563362950 Chr.2: 97725185-97725185 on Build GRCh38
rs777285124 Chr.2: 97725121-97725121 on Build GRCh38
rs773287941 Chr.2: 97725167-97725167 on Build GRCh38
rs867822093 Chr.2: 97725103-97725103 on Build GRCh38
rs199864362 Chr.2: 97725094-97725094 on Build GRCh38
rs750301906 Chr.2: 97725075-97725075 on Build GRCh38
rs201206711 Chr.2: 97725116-97725116 on Build GRCh38
rs145955907 Chr.2: 97725153-97725153 on Build GRCh38
rs764657700 Chr.2: 97725209-97725209 on Build GRCh38
rs766233841 Chr.2: 97725196-97725196 on Build GRCh38
rs960299379 Chr.2: 97725241-97725241 on Build GRCh38
rs759485019 Chr.2: 97725208-97725208 on Build GRCh38
rs202193449 Chr.2: 97725089-97725089 on Build GRCh38
rs199742344 Chr.2: 97725154-97725154 on Build GRCh38
rs777528648 Chr.2: 97725216-97725216 on Build GRCh38
rs2289918 Chr.2: 97725265-97725265 on Build GRCh38
rs201040139 Chr.2: 97725276-97725277 on Build GRCh38
rs55964305 Chr.2: 97725213-97725213 on Build GRCh38
rs200552725 Chr.2: 97725157-97725157 on Build GRCh38
rs754563357 Chr.2: 97725098-97725098 on Build GRCh38
rs752318274 Chr.2: 97725212-97725212 on Build GRCh38
rs868238887 Chr.2: 97725081-97725081 on Build GRCh38

View Full Product Details