Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32439850-32440123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRA
Entrez Gene ID
3122
Gene Name
major histocompatibility complex, class II, DR alpha
Gene Aliases
HLA-DRA1, MLRW
UniGene
Hs.520048
Gene Chromosome Location
Chr.6: 32439842-32445046 on Build GRCh38
Transcripts
NM_019111.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1312056 c.19C>T p.P7S Chr.6: 32439969-32439969 on Build GRCh38
6358266 c.82+9T>C p.? Chr.6: 32440041-32440041 on Build GRCh38
4610506 c.82G>A p.E28K Chr.6: 32440032-32440032 on Build GRCh38
5928815 c.11G>A p.S4N Chr.6: 32439961-32439961 on Build GRCh38
7106955 c.70T>C p.W24R Chr.6: 32440020-32440020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771937138 Chr.6: 32440018-32440018 on Build GRCh38
rs267600976 Chr.6: 32439953-32439953 on Build GRCh38
rs757574147 Chr.6: 32439987-32439987 on Build GRCh38
rs772938215 Chr.6: 32440019-32440019 on Build GRCh38
rs200899094 Chr.6: 32439933-32439933 on Build GRCh38
rs764188346 Chr.6: 32440050-32440050 on Build GRCh38
rs146349593 Chr.6: 32440072-32440072 on Build GRCh38
rs41270484 Chr.6: 32440039-32440039 on Build GRCh38
rs760658180 Chr.6: 32440026-32440026 on Build GRCh38
rs762033414 Chr.6: 32440062-32440062 on Build GRCh38
rs747809079 Chr.6: 32440014-32440014 on Build GRCh38
rs199928204 Chr.6: 32439978-32439978 on Build GRCh38
rs762969595 Chr.6: 32440038-32440038 on Build GRCh38
rs781412925 Chr.6: 32439989-32439989 on Build GRCh38
rs950252086 Chr.6: 32439928-32439928 on Build GRCh38
rs749337257 Chr.6: 32439944-32439945 on Build GRCh38
rs367898223 Chr.6: 32439925-32439925 on Build GRCh38
rs756633767 Chr.6: 32439992-32439992 on Build GRCh38
rs201021461 Chr.6: 32439939-32439939 on Build GRCh38
rs370932716 Chr.6: 32439908-32439908 on Build GRCh38
rs367604927 Chr.6: 32439940-32439940 on Build GRCh38
rs14004 Chr.6: 32439932-32439932 on Build GRCh38
rs770813718 Chr.6: 32440027-32440027 on Build GRCh38
rs768468269 Chr.6: 32439919-32439919 on Build GRCh38
rs951021159 Chr.6: 32439878-32439878 on Build GRCh38
rs891176272 Chr.6: 32440044-32440044 on Build GRCh38
rs766751012 Chr.6: 32440075-32440075 on Build GRCh38
rs765347014 Chr.6: 32439954-32439954 on Build GRCh38
rs142154804 Chr.6: 32440008-32440008 on Build GRCh38
rs143284297 Chr.6: 32440004-32440004 on Build GRCh38
rs371443293 Chr.6: 32440007-32440007 on Build GRCh38
rs542202057 Chr.6: 32440023-32440023 on Build GRCh38
rs763131341 Chr.6: 32439957-32439957 on Build GRCh38
rs775617318 Chr.6: 32440030-32440030 on Build GRCh38
rs138394438 Chr.6: 32439963-32439963 on Build GRCh38
rs750816033 Chr.6: 32440071-32440071 on Build GRCh38
rs755760382 Chr.6: 32439958-32439958 on Build GRCh38
rs781626637 Chr.6: 32439910-32439910 on Build GRCh38
rs754430579 Chr.6: 32440077-32440077 on Build GRCh38
rs752785871 Chr.6: 32439955-32439955 on Build GRCh38
rs751711070 Chr.6: 32440061-32440061 on Build GRCh38
rs549063239 Chr.6: 32439937-32439937 on Build GRCh38
rs16822586 Chr.6: 32439996-32439996 on Build GRCh38
rs769278344 Chr.6: 32439912-32439912 on Build GRCh38
rs367986407 Chr.6: 32439931-32439931 on Build GRCh38
rs750880721 Chr.6: 32439990-32439990 on Build GRCh38
rs1018549744 Chr.6: 32440076-32440076 on Build GRCh38
rs16822587 Chr.6: 32440078-32440078 on Build GRCh38
rs73407183 Chr.6: 32439911-32439911 on Build GRCh38
rs767906653 Chr.6: 32440064-32440064 on Build GRCh38
rs879152274 Chr.6: 32440032-32440033 on Build GRCh38
rs371818250 Chr.6: 32440094-32440094 on Build GRCh38
rs748952885 Chr.6: 32439917-32439917 on Build GRCh38

View Full Product Details