Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154546652-154546885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PD
Entrez Gene ID
2539
Gene Name
glucose-6-phosphate dehydrogenase
Gene Aliases
G6PD1
UniGene
Hs.461047
Gene Chromosome Location
Chr.X: 154531390-154547586 on Build GRCh38
Transcripts
NM_000402.4 NM_001042351.2 NM_001099856.4 NM_001321396.1

Gene Symbol
IKBKG
Entrez Gene ID
8517
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
Gene Aliases
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9
UniGene
Hs.43505
Gene Chromosome Location
Chr.X: 154542240-154565046 on Build GRCh38
Transcripts
NM_000402.4 NM_001042351.2 NM_001099856.4 NM_001321396.1

SNP Detail

SNP ID Chromosome Location
rs781989843 Chr.X: 154546782-154546782 on Build GRCh38
rs782006658 Chr.X: 154546800-154546800 on Build GRCh38
rs782644091 Chr.X: 154546758-154546759 on Build GRCh38
rs782023794 Chr.X: 154546803-154546803 on Build GRCh38
rs782493718 Chr.X: 154546859-154546859 on Build GRCh38
rs782562909 Chr.X: 154546753-154546753 on Build GRCh38
rs368832112 Chr.X: 154546794-154546794 on Build GRCh38
rs782230848 Chr.X: 154546847-154546847 on Build GRCh38
rs782700083 Chr.X: 154546823-154546823 on Build GRCh38
rs782150651 Chr.X: 154546851-154546851 on Build GRCh38
rs782232679 Chr.X: 154546783-154546783 on Build GRCh38
rs782375017 Chr.X: 154546744-154546744 on Build GRCh38
rs782199182 Chr.X: 154546863-154546863 on Build GRCh38
rs782307351 Chr.X: 154546865-154546865 on Build GRCh38
rs782539588 Chr.X: 154546847-154546847 on Build GRCh38
rs934195145 Chr.X: 154546713-154546713 on Build GRCh38
rs782318674 Chr.X: 154546742-154546742 on Build GRCh38
rs782758722 Chr.X: 154546812-154546812 on Build GRCh38
rs921512630 Chr.X: 154546709-154546709 on Build GRCh38
rs782344302 Chr.X: 154546784-154546784 on Build GRCh38
rs782130452 Chr.X: 154546804-154546804 on Build GRCh38
rs781940446 Chr.X: 154546788-154546788 on Build GRCh38
rs1052584224 Chr.X: 154546791-154546791 on Build GRCh38
rs781837306 Chr.X: 154546816-154546816 on Build GRCh38
rs782395004 Chr.X: 154546780-154546780 on Build GRCh38
rs782663603 Chr.X: 154546849-154546849 on Build GRCh38
rs935099185 Chr.X: 154546779-154546779 on Build GRCh38
rs782481467 Chr.X: 154546817-154546817 on Build GRCh38
rs782291173 Chr.X: 154546766-154546766 on Build GRCh38
rs986916864 Chr.X: 154546752-154546752 on Build GRCh38
rs782609860 Chr.X: 154546862-154546862 on Build GRCh38
rs911665034 Chr.X: 154546755-154546755 on Build GRCh38
rs781784247 Chr.X: 154546837-154546837 on Build GRCh38

View Full Product Details