Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32443268-32443501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRA
Entrez Gene ID
3122
Gene Name
major histocompatibility complex, class II, DR alpha
Gene Aliases
HLA-DRA1, MLRW
UniGene
Hs.520048
Gene Chromosome Location
Chr.6: 32439842-32445046 on Build GRCh38
Transcripts
NM_019111.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5034994 c.512G>A p.R171H Chr.6: 32443368-32443368 on Build GRCh38
3348734 c.435G>A p.T145T Chr.6: 32443291-32443291 on Build GRCh38
4901056 c.580G>A p.G194S Chr.6: 32443436-32443436 on Build GRCh38
4997786 c.568G>T p.V190L Chr.6: 32443424-32443424 on Build GRCh38
4831184 c.595C>A p.L199I Chr.6: 32443451-32443451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532040678 Chr.6: 32443291-32443291 on Build GRCh38
rs372692651 Chr.6: 32443431-32443431 on Build GRCh38
rs879138868 Chr.6: 32443469-32443469 on Build GRCh38
rs146370028 Chr.6: 32443388-32443388 on Build GRCh38
rs770825671 Chr.6: 32443415-32443415 on Build GRCh38
rs200798853 Chr.6: 32443409-32443409 on Build GRCh38
rs142014572 Chr.6: 32443360-32443360 on Build GRCh38
rs752606047 Chr.6: 32443414-32443414 on Build GRCh38
rs936299247 Chr.6: 32443312-32443312 on Build GRCh38
rs769243656 Chr.6: 32443320-32443320 on Build GRCh38
rs763361772 Chr.6: 32443317-32443317 on Build GRCh38
rs559274633 Chr.6: 32443332-32443332 on Build GRCh38
rs760381879 Chr.6: 32443381-32443381 on Build GRCh38
rs946681808 Chr.6: 32443290-32443290 on Build GRCh38
rs867285682 Chr.6: 32443298-32443298 on Build GRCh38
rs199723815 Chr.6: 32443433-32443433 on Build GRCh38
rs750128905 Chr.6: 32443349-32443349 on Build GRCh38
rs537246240 Chr.6: 32443470-32443470 on Build GRCh38
rs753708036 Chr.6: 32443408-32443408 on Build GRCh38
rs773709185 Chr.6: 32443326-32443326 on Build GRCh38
rs780246792 Chr.6: 32443451-32443451 on Build GRCh38
rs138462914 Chr.6: 32443299-32443299 on Build GRCh38
rs756467266 Chr.6: 32443447-32443447 on Build GRCh38
rs997782030 Chr.6: 32443392-32443392 on Build GRCh38
rs772798906 Chr.6: 32443367-32443367 on Build GRCh38
rs749707818 Chr.6: 32443461-32443461 on Build GRCh38
rs552360101 Chr.6: 32443472-32443472 on Build GRCh38

View Full Product Details