Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97734343-97734616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
UniGene
Hs.234569
Gene Chromosome Location
Chr.2: 97712030-97744327 on Build GRCh38
Transcripts
NM_001079.3 NM_207519.1 XM_017004867.1 XM_017004868.1 XM_017004869.1 XM_017004870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5896680 c.542C>T p.P181L Chr.2: 97734550-97734550 on Build GRCh38
1270671 c.935C>T p.T312M Chr.2: 97734565-97734565 on Build GRCh38
5896682 c.541C>T p.P181S Chr.2: 97734549-97734549 on Build GRCh38
73325 c.908A>G p.D303G Chr.2: 97734538-97734538 on Build GRCh38
6824749 c.522G>A p.T174T Chr.2: 97734530-97734530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566698608 Chr.2: 97734373-97734373 on Build GRCh38
rs200679935 Chr.2: 97734570-97734570 on Build GRCh38
rs538552532 Chr.2: 97734384-97734384 on Build GRCh38
rs568821265 Chr.2: 97734423-97734423 on Build GRCh38
rs1001220096 Chr.2: 97734365-97734365 on Build GRCh38
rs199993140 Chr.2: 97734591-97734591 on Build GRCh38
rs770861777 Chr.2: 97734576-97734576 on Build GRCh38
rs762428344 Chr.2: 97734544-97734544 on Build GRCh38
rs148202842 Chr.2: 97734398-97734398 on Build GRCh38
rs201552454 Chr.2: 97734584-97734584 on Build GRCh38
rs145218891 Chr.2: 97734569-97734569 on Build GRCh38
rs770806735 Chr.2: 97734495-97734495 on Build GRCh38
rs779465321 Chr.2: 97734566-97734566 on Build GRCh38
rs778114683 Chr.2: 97734486-97734486 on Build GRCh38
rs964798214 Chr.2: 97734496-97734496 on Build GRCh38
rs1025521945 Chr.2: 97734421-97734421 on Build GRCh38
rs778794433 Chr.2: 97734469-97734469 on Build GRCh38
rs772485269 Chr.2: 97734485-97734485 on Build GRCh38
rs955373496 Chr.2: 97734422-97734422 on Build GRCh38
rs763568575 Chr.2: 97734455-97734455 on Build GRCh38
rs201386206 Chr.2: 97734550-97734550 on Build GRCh38
rs752096371 Chr.2: 97734535-97734538 on Build GRCh38
rs754376652 Chr.2: 97734551-97734551 on Build GRCh38
rs191682943 Chr.2: 97734504-97734504 on Build GRCh38
rs1022235755 Chr.2: 97734563-97734563 on Build GRCh38
rs199612427 Chr.2: 97734505-97734505 on Build GRCh38
rs200510184 Chr.2: 97734546-97734546 on Build GRCh38
rs374461803 Chr.2: 97734492-97734492 on Build GRCh38
rs200214533 Chr.2: 97734522-97734522 on Build GRCh38
rs903840321 Chr.2: 97734565-97734565 on Build GRCh38
rs201560822 Chr.2: 97734523-97734523 on Build GRCh38
rs891705207 Chr.2: 97734478-97734483 on Build GRCh38
rs1053362907 Chr.2: 97734449-97734450 on Build GRCh38
rs938523452 Chr.2: 97734445-97734445 on Build GRCh38
rs574174678 Chr.2: 97734535-97734535 on Build GRCh38
rs200862014 Chr.2: 97734548-97734548 on Build GRCh38
rs141311297 Chr.2: 97734457-97734457 on Build GRCh38
rs745670298 Chr.2: 97734590-97734590 on Build GRCh38
rs765996782 Chr.2: 97734518-97734519 on Build GRCh38
rs973731366 Chr.2: 97734387-97734387 on Build GRCh38
rs759332713 Chr.2: 97734509-97734509 on Build GRCh38
rs748259677 Chr.2: 97734480-97734480 on Build GRCh38
rs769350009 Chr.2: 97734529-97734529 on Build GRCh38
rs879170480 Chr.2: 97734493-97734493 on Build GRCh38
rs199705818 Chr.2: 97734530-97734530 on Build GRCh38
rs906983231 Chr.2: 97734427-97734427 on Build GRCh38

View Full Product Details