Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97734561-97734814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
UniGene
Hs.234569
Gene Chromosome Location
Chr.2: 97712030-97744327 on Build GRCh38
Transcripts
NM_001079.3 NM_207519.1 XM_017004867.1 XM_017004868.1 XM_017004869.1 XM_017004870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7213106 c.700C>T p.R234C Chr.2: 97734708-97734708 on Build GRCh38
6509590 c.687C>A p.G229G Chr.2: 97734695-97734695 on Build GRCh38
3729200 c.1036T>C p.C346R Chr.2: 97734666-97734666 on Build GRCh38
5059132 c.1065C>T p.G355G Chr.2: 97734695-97734695 on Build GRCh38
1270671 c.935C>T p.T312M Chr.2: 97734565-97734565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008492987 Chr.2: 97734786-97734786 on Build GRCh38
rs200119693 Chr.2: 97734631-97734631 on Build GRCh38
rs771668086 Chr.2: 97734732-97734732 on Build GRCh38
rs532293567 Chr.2: 97734787-97734787 on Build GRCh38
rs753212430 Chr.2: 97734665-97734665 on Build GRCh38
rs767197242 Chr.2: 97734636-97734636 on Build GRCh38
rs955295414 Chr.2: 97734784-97734784 on Build GRCh38
rs754217622 Chr.2: 97734639-97734639 on Build GRCh38
rs777939833 Chr.2: 97734689-97734689 on Build GRCh38
rs201318076 Chr.2: 97734722-97734722 on Build GRCh38
rs371574765 Chr.2: 97734618-97734618 on Build GRCh38
rs757486529 Chr.2: 97734705-97734705 on Build GRCh38
rs774008070 Chr.2: 97734624-97734624 on Build GRCh38
rs751054040 Chr.2: 97734794-97734794 on Build GRCh38
rs1016209671 Chr.2: 97734629-97734629 on Build GRCh38
rs869025224 Chr.2: 97734709-97734709 on Build GRCh38
rs746814529 Chr.2: 97734789-97734789 on Build GRCh38
rs780358888 Chr.2: 97734774-97734774 on Build GRCh38
rs200537559 Chr.2: 97734730-97734730 on Build GRCh38
rs200487627 Chr.2: 97734744-97734744 on Build GRCh38
rs199863426 Chr.2: 97734657-97734657 on Build GRCh38
rs768157128 Chr.2: 97734729-97734729 on Build GRCh38
rs758571157 Chr.2: 97734668-97734668 on Build GRCh38
rs200511463 Chr.2: 97734603-97734603 on Build GRCh38
rs745670298 Chr.2: 97734590-97734590 on Build GRCh38
rs759853845 Chr.2: 97734739-97734739 on Build GRCh38
rs201620644 Chr.2: 97734742-97734742 on Build GRCh38
rs55764292 Chr.2: 97734757-97734757 on Build GRCh38
rs55933862 Chr.2: 97734720-97734720 on Build GRCh38
rs781392738 Chr.2: 97734750-97734751 on Build GRCh38
rs755585715 Chr.2: 97734718-97734726 on Build GRCh38
rs764695602 Chr.2: 97734752-97734752 on Build GRCh38
rs772894578 Chr.2: 97734733-97734733 on Build GRCh38
rs768529454 Chr.2: 97734793-97734793 on Build GRCh38
rs201207626 Chr.2: 97734695-97734695 on Build GRCh38
rs947259720 Chr.2: 97734755-97734755 on Build GRCh38
rs920192176 Chr.2: 97734659-97734659 on Build GRCh38
rs199993140 Chr.2: 97734591-97734591 on Build GRCh38
rs199629898 Chr.2: 97734743-97734743 on Build GRCh38
rs201221859 Chr.2: 97734642-97734642 on Build GRCh38
rs763489646 Chr.2: 97734751-97734751 on Build GRCh38
rs201552454 Chr.2: 97734584-97734584 on Build GRCh38
rs1052946031 Chr.2: 97734750-97734750 on Build GRCh38
rs202131881 Chr.2: 97734702-97734702 on Build GRCh38
rs200318575 Chr.2: 97734747-97734747 on Build GRCh38
rs201605654 Chr.2: 97734611-97734611 on Build GRCh38
rs542219545 Chr.2: 97734721-97734721 on Build GRCh38
rs948337308 Chr.2: 97734666-97734666 on Build GRCh38
rs761391482 Chr.2: 97734632-97734632 on Build GRCh38
rs769677892 Chr.2: 97734599-97734599 on Build GRCh38
rs113310375 Chr.2: 97734697-97734697 on Build GRCh38
rs142702703 Chr.2: 97734655-97734655 on Build GRCh38
rs570859815 Chr.2: 97734775-97734775 on Build GRCh38
rs781463009 Chr.2: 97734708-97734708 on Build GRCh38
rs900434958 Chr.2: 97734781-97734781 on Build GRCh38
rs201365297 Chr.2: 97734728-97734728 on Build GRCh38

View Full Product Details