Gene Transcripts

Gene Symbol
Nfs1
Entrez Gene ID
18041
Gene Name
nitrogen fixation gene 1 (S. cerevisiae)
Gene Aliases
AA987187, m-Nfs1, m-Nfsl
Chromosome Location
Chr. 2: 156123637 - 156144186 on Build GRCm38
UniGene ID
Mm.283604
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010911.2 NP_035041.2 5 476
GenBank mRNA AJ222660.1 - 5 475
AK005724.1 - 5 465
AK075575.1 - 5 474
AK132838.1 - 5 477
AK148527.1 - 5 477
AK159828.1 - 5 465
BC010586.1 - 5 454 3711352

View Full Product Details