Gene Transcripts

Gene Symbol
Tmprss15
Entrez Gene ID
19146
Gene Name
transmembrane protease, serine 15
Gene Aliases
Entk, Prss7
Chromosome Location
Chr. 16: 78953008 - 79091097 on Build GRCm38
UniGene ID
Mm.5184
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008941.3 NP_032967.1 12 1320
NM_178855.4 NP_849186.2 11 1275
GenBank mRNA AK033582.1 - 11 1277
AK038356.1 - 3 269
BC117917.1 - 12 1319 40097367
BC117918.1 - 12 1319 40097370
U73378.1 - 12 1315

View Full Product Details