Gene Transcripts

Gene Symbol
Plekhs1
Entrez Gene ID
226245
Gene Name
pleckstrin homology domain containing, family S member 1
Gene Aliases
9930023K05Rik
Chromosome Location
Chr. 19: 56457276 - 56486729 on Build GRCm38
UniGene ID
Mm.156742
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164263.1 NP_001157735.1 4 248
NM_172641.3 NP_766229.2 4 248
GenBank mRNA AK036908.1 - 4 246
AK053921.1 - 4 246
BC090250.1 - 4 226 6485933

View Full Product Details