Gene Transcripts
- Gene Symbol
- Olfr1009
- Entrez Gene ID
- 258565
- Gene Name
- olfactory receptor 1009
- Gene Aliases
- MOR175-3
- Chromosome Location
- Chr. 2: 85721407 - 85722351 on Build GRCm38
- UniGene ID
- Mm.377570
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC111578.1 |
- |
1 |
252 |
|
BC148238.1 |
- |
1 |
253 |
40042413 |
View Full Product Details