Gene Transcripts

Gene Symbol
Npsr1
Entrez Gene ID
319239
Gene Name
neuropeptide S receptor 1
Gene Aliases
9330128H10Rik, GPRA, Gpr154, MVTR, PGR14, VRR1
Chromosome Location
Chr. 9: 24097950 - 24317590 on Build GRCm38
UniGene ID
Mm.130824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175678.3 NP_783609.1 6, 7 764
GenBank mRNA AK033957.1 - 6, 7 765
AK041451.1 - 5, 6 663
AY299387.1 - 5, 6 700

View Full Product Details