Gene Transcripts

Gene Symbol
Adgrf1
Entrez Gene ID
77596
Gene Name
adhesion G protein-coupled receptor F1
Gene Aliases
5031409J19Rik, Gpr110, KPG_009
Chromosome Location
Chr. 17: 43270276 - 43325876 on Build GRCm38
UniGene ID
Mm.159950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133776.2 NP_598537.2 12 2327
GenBank mRNA AK019869.1 - 12 2331
BC019217.1 - 12 2356 5065702

View Full Product Details