Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030026.2 | NP_084302.1 | 6 | 711 | |
GenBank mRNA | AK132265.1 | - | 6 | 691 | |
AK143233.1 | - | 6 | 712 | ||
AK145564.1 | - | 6 | 663 | ||
AK146844.1 | - | 6 | 671 | ||
AK146865.1 | - | 6 | 661 | ||
BC141193.1 | - | 6 | 677 | 9055805 | |
BC141194.1 | - | 6 | 677 | 9055806 | |
BQ551658.1 | - | 2 | 163 |