Gene Transcripts
- Gene Symbol
- Lyn
- Entrez Gene ID
- 17096
- Gene Name
- Yamaguchi sarcoma viral (v-yes-1) oncogene homolog
- Gene Aliases
- AA407514, Hck-2, p53Lyn, p56Lyn
- Chromosome Location
- Chr. 4: 3678121 - 3791612 on Build GRCm38
- UniGene ID
- Mm.317331
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001111096.1 |
NP_001104566.1
|
11, 12 |
1458 |
|
NM_010747.2 |
NP_034877.2
|
11, 12 |
1395 |
|
GenBank mRNA |
AK028112.1 |
- |
11, 12 |
1391 |
|
AK151914.1 |
- |
11, 12 |
1454 |
|
AK152199.1 |
- |
11, 12 |
1456 |
|
AK153038.1 |
- |
11, 12 |
1392 |
|
AK170561.1 |
- |
11, 12 |
1456 |
|
BC003253.1 |
- |
11, 12 |
1335 |
3500419 |
BC031547.1 |
- |
11, 12 |
1428 |
3589207 |
BC157998.1 |
- |
11, 12 |
1227 |
9007404 |
M33426.1 |
- |
1, 2 |
122 |
|
M57696.1 |
- |
11, 12 |
1294 |
|
M57697.1 |
- |
11, 12 |
1231 |
|
M64608.1 |
- |
11, 12 |
1444 |
|
View Full Product Details