Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297606.1 | NP_001284535.1 | 1 | 146 | |
NM_001297607.1 | NP_001284536.1 | 1 | 146 | ||
NM_009010.5 | NP_033036.2 | 1 | 146 | ||
GenBank mRNA | AK009733.1 | - | 1 | 123 | |
AK038300.1 | - | 1 | 123 | ||
AK045224.1 | - | 1 | 136 | ||
AK161875.1 | - | 1 | 123 | ||
AK165467.1 | - | 1 | 147 | ||
BC003846.1 | - | 1 | 134 | ||
BC138684.1 | - | 1 | 106 | 8861705 | |
BC145372.1 | - | 1 | 106 | 9053930 |