Gene Transcripts
- Gene Symbol
- Olfr1
- Entrez Gene ID
- 258923
- Gene Name
- olfactory receptor 1
- Gene Aliases
- I54, MOR135-13
- Chromosome Location
- Chr. 11: 73395076 - 73399495 on Build GRCm38
- UniGene ID
- Mm.246523
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146921.2 |
NP_667132.1
|
3 |
997 |
|
View Full Product Details